Print…

Anti-SEPT12 Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBI041560EA 587 EUR
USBI041560
Anti-SEPT12 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SEPT12 (septin 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR