Print…
Anti-SEPT11 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornecedor: United States Biological
USBI041559-PEEA 852 EUR
USBI041559-PE
Anti-SEPT11 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT11 (septin 11)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human