Print…
Anti-SEPT11 Rabbit Polyclonal Antibody (Biotin)
Fornecedor: United States Biological
USBI041559-BIOTINEA 852 EUR
USBI041559-BIOTIN
Anti-SEPT11 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPT11 (septin 11)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human