Print…

Anti-SEPSECS Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBI041556EA 587 EUR
USBI041556
Anti-SEPSECS Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SEPSECS (Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR