Print…
Anti-SEPP1 Rabbit Polyclonal Antibody (Biotin)
Fornecedor: United States Biological
USBI041555-BIOTINEA 852 EUR
USBI041555-BIOTIN
Anti-SEPP1 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPP1 (selenoprotein P, plasma, 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human