Print…
Anti-SEPP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornecedor: United States Biological
USBI041555-APCEA 852 EUR
USBI041555-APC
Anti-SEPP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPP1 (selenoprotein P, plasma, 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human