Print…
Anti-SENP6 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: United States Biological
USBI041552-APEA 852 EUR
USBI041552-AP
Anti-SENP6 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence
Do not freeze!
Type: Primary
Antigen: SENP6 (SUMO1/sentrin specific peptidase 6)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human