Print…

Anti-SENP6 Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBI041552EA 587 EUR
USBI041552
Anti-SENP6 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence

Type: Primary
Antigen: SENP6 (SUMO1/sentrin specific peptidase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR