Print…
Anti-HIRA Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: US Biological
USBI036572-APEA 852 EUR
USBI036572-AP
Anti-HIRA Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HIRA (histone cell cycle regulator)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human