Print…
Anti-HIPK4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: US Biological
USBI036570-HRPEA 852 EUR
USBI036570-HRP
Anti-HIPK4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: HIPK4 (homeodomain interacting protein kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human