Print…
Anti-HIPK4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornecedor: US Biological
USBI036570-FITCEA 852 EUR
USBI036570-FITC
Anti-HIPK4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: HIPK4 (homeodomain interacting protein kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human