Print…
Anti-HIPK3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornecedor: US Biological
USBI036568-PEEA 852 EUR
USBI036568-PE
Anti-HIPK3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: HIPK3 (homeodomain interacting protein kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human