Print…

Anti-HIPK1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornecedor: US Biological
USBI036563-FITCEA 852 EUR
USBI036563-FITC
Anti-HIPK1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: HIPK1 (homeodomain interacting protein kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR