Print…
Anti-HIPK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: US Biological
USBI036563-APEA 852 EUR
USBI036563-AP
Anti-HIPK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: HIPK1 (homeodomain interacting protein kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human