Print…
Anti-HIP1R Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: US Biological
USBI036562-HRPEA 852 EUR
USBI036562-HRP
Anti-HIP1R Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: HIP1R (huntingtin interacting protein 1 related)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human