Print…

Anti-HINFP Rabbit Polyclonal Antibody (Biotin)

Fornecedor: US Biological
USBI036558-BIOTINEA 852 EUR
USBI036558-BIOTIN
Anti-HINFP Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HINFP (histone H4 transcription factor)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR