Print…
Anti-HINFP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: US Biological
USBI036558-APEA 852 EUR
USBI036558-AP
Anti-HINFP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HINFP (histone H4 transcription factor)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human