Print…
Anti-HILS1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: US Biological
USBI036557-APEA 852 EUR
USBI036557-AP
Anti-HILS1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HILS1 (histone linker H1 domain, spermatid-specific 1, pseudogene)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human