Print…
Anti-HIGD2A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: US Biological
USBI036556-HRPEA 852 EUR
USBI036556-HRP
Anti-HIGD2A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: HIGD2A (HIG1 hypoxia inducible domain family, member 2A)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human