Print…
Anti-HIGD1C Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornecedor: US Biological
USBI036555-PEEA 852 EUR
USBI036555-PE
Anti-HIGD1C Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HIGD1C (HIG1 hypoxia inducible domain family, member 1C)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse