Print…

Anti-CAMK2N1 Rabbit Polyclonal Antibody (Biotin)

Fornecedor: United States Biological
USBI033164-BIOTINEA 852 EUR
USBI033164-BIOTIN
Anti-CAMK2N1 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CAMK2N1 (calcium/calmodulin-dependent protein kinase II inhibitor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR