Print…
Anti-CAMK2D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornecedor: United States Biological
USBI033162-FITCEA 852 EUR
USBI033162-FITC
Anti-CAMK2D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CAMK2D (calcium/calmodulin-dependent protein kinase II delta)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse