Print…

Anti-CAMK2D Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornecedor: United States Biological
USBI033162-APEA 852 EUR
USBI033162-AP
Anti-CAMK2D Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CAMK2D (calcium/calmodulin-dependent protein kinase II delta)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR