Print…

Anti-CAMK2B Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornecedor: United States Biological
USBI033161-PEEA 852 EUR
USBI033161-PE
Anti-CAMK2B Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CAMK2B (calcium/calmodulin-dependent protein kinase II beta)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR