Print…

Anti-NUP50 Mouse Monoclonal Antibody [clone: 247CT10.2.5]

Fornecedor: United States Biological
USBI031170EA 487 EUR
USBI031170
Anti-NUP50 Mouse Monoclonal Antibody [clone: 247CT10.2.5]
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1560

Type: Primary
Antigen: NUP50 (nucleoporin 50kDa)
Clonality: Monoclonal
Clone: 247CT10.2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR