Print…

Anti-NUCL Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 130CT43.8.5]

Fornecedor: United States Biological
USBI031168-APEA 852 EUR
USBI031168-AP
Anti-NUCL Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 130CT43.8.5]
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: NUCL
Clonality: Monoclonal
Clone: 130CT43.8.5
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR