Print…
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 185CT47.6.1]
Fornecedor: United States Biological
USBI031165EA 577 EUR
USBI031165
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 185CT47.6.1]
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-4000
Type: Primary
Antigen: NTRK1 (neurotrophic tyrosine kinase, receptor, type 1)
Clonality: Monoclonal
Clone: 185CT47.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse