Print…

Anti-NPM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3A9F1]

Fornecedor: United States Biological
USBI031163-PEEA 616 EUR
USBI031163-PE
Anti-NPM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3A9F1]
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:460
Do not freeze! Light sensitive

Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: Monoclonal
Clone: 3A9F1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR