Print…

Anti-NOTCH3 Mouse Monoclonal Antibody [clone: 487CT6.9.2]

Fornecedor: United States Biological
USBI031162EA 587 EUR
USBI031162
Anti-NOTCH3 Mouse Monoclonal Antibody [clone: 487CT6.9.2]
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality: Monoclonal
Clone: 487CT6.9.2
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR