Print…

Anti-NANOG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 60CT77.1.1]

Fornecedor: United States Biological
USBI031151-FITCEA 711 EUR
USBI031151-FITC
Anti-NANOG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 60CT77.1.1]
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250; Immunochemistry: 1:10-50

Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: Monoclonal
Clone: 60CT77.1.1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR