Print…

Anti-MB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 444CT19.2.1]

Fornecedor: United States Biological
USBI031150-PEEA 852 EUR
USBI031150-PE
Anti-MB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 444CT19.2.1]
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250-1000
Do not freeze! Light sensitive

Type: Primary
Antigen: MB (myoglobin)
Clonality: Monoclonal
Clone: 444CT19.2.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR