Print…

Anti-MYOG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 304CT11.6.1]

Fornecedor: United States Biological
USBI031148-PEEA 852 EUR
USBI031148-PE
Anti-MYOG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 304CT11.6.1]
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100
Do not freeze! Light sensitive

Type: Primary
Antigen: MYOG (myogenin (myogenic factor 4))
Clonality: Monoclonal
Clone: 304CT11.6.1
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR