Print…

Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Fornecedor: Rockland Immunochemicals
ROCK710-106-130EA 650 EUR
ROCK710-106-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Anticorpos
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Recommended Dilutions: ELISA: 1:1200,000; Western Blot: 1:5000 - 1:20000; Immunochemistry: 1:2000 - 1:20000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR