Print…

Anti-AIRE Goat Polyclonal Antibody

Fornecedor: Everest Biotechnology
EVBIEB05507EA 325.5 EUR
EVBIEB05507
Anti-AIRE Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR