Print…
Anti-IgG2b Goat Polyclonal Antibody (CF™ 647)
Fornecedor: Biotium
20272.EA 372 EUR
20272. 20272-1
Anti-IgG2b Goat Polyclonal Antibody (CF™ 647)
Anticorpos
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse