Print…
Anti-PEPD Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111851-100EA 993 EUR
ABCAAB111851-100
Anti-PEPD Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PEPD (peptidase D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human