Print…
Anti-FUZ Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111842-50EA 921 EUR
ABCAAB111842-50
Anti-FUZ Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: FUZ (fuzzy planar cell polarity protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human