Print…
Anti-RASGRF1 Rabbit Monoclonal Antibody [clone: EPR1790]
Fornecedor: Abcam
ABCAAB111830-100EA 908 EUR
ABCAAB111830-100
Anti-RASGRF1 Rabbit Monoclonal Antibody [clone: EPR1790]
Anticorpos
Type: Primary
Antigen: RASGRF1 (Ras protein-specific guanine nucleotide-releasing factor 1)
Clonality: monoclonal
Clone: EPR1790
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar