Print…

Anti-TPH2 Rabbit Polyclonal Antibody

Fornecedor: Abcam
ABCAAB111828-100EA 984 EUR
ABCAAB111828-100
Anti-TPH2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR