Print…
Anti-PAR6 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111823-100EA 759 EUR
ABCAAB111823-100
Anti-PAR6 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PAR6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human