Print…
Anti-WEE1 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111820-100EA 971 EUR
ABCAAB111820-100
Anti-WEE1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: WEE1 (WEE1 G2 checkpoint kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human