Print…
Anti-PAX1 Rabbit polyclonal antibody unconjugated
ABCAAB111752-400EA 751 EUR
ABCAAB111752-400
Anti-PAX1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: PAX1 (paired box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse