Print…
Anti-CES2 Rabbit polyclonal antibody unconjugated
ABCAAB111751-400EA 740 EUR
ABCAAB111751-400
Anti-CES2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CES2 (carboxylesterase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human