Print…
Anti-NRF1 Rabbit polyclonal antibody unconjugated
ABCAAB111746-100EA 740 EUR
ABCAAB111746-100
Anti-NRF1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: NRF1 (nuclear respiratory factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human