Print…
Anti-COL4A3 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111742-100EA 965 EUR
ABCAAB111742-100
Anti-COL4A3 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: COL4A3 (collagen, type IV, alpha 3 (Goodpasture antigen))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human