Print…
Anti-VCP Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111740-50EA 1020 EUR
ABCAAB111740-50
Anti-VCP Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: VCP (valosin containing protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human