Print…
Anti-KRT31 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111730-50EA 921 EUR
ABCAAB111730-50
Anti-KRT31 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: KRT31 (keratin 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human