Print…
Anti-Elav-type RNA-binding protein ETR3 Rabbit polyclonal antibody unconjugated
Fornecedor: Abcam
ABCAAB111728-50EA 774 EUR
ABCAAB111728-50
Anti-Elav-type RNA-binding protein ETR3 Rabbit polyclonal antibody unconjugated
Anticorpos
Type: Primary
Antigen: Elav-type RNA-binding protein ETR3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human