Print…
Anti-PTN Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111725-100EA 944 EUR
ABCAAB111725-100
Anti-PTN Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PTN (pleiotrophin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human