Print…
Anti-BCAN Rabbit polyclonal antibody unconjugated
Fornecedor: Abcam
ABCAAB111719-50EA 974 EUR
ABCAAB111719-50
Anti-BCAN Rabbit polyclonal antibody unconjugated
Anticorpos
Type: Primary
Antigen: BCAN (brevican)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human